Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fktn
Entrez Gene ID
246179
Gene Name
fukutin
Gene Aliases
D830030O17Rik, Fcmd
Chromosome Location
Chr.4: 53714107 - 53765785 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB077383.1 - 1 - 2 194 109

View Full Product Details