The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178061.5 | NP_835162.1 | 2 - 3 | 853 | 110 |
GenBank mRNA | AK039841.1 | - | 2 - 3 | 699 | 110 |
AK155217.1 | - | 2 - 3 | 854 | 110 | |
AK170973.1 | - | 2 - 3 | 847 | 110 | |
BC020028.1 | - | 2 - 3 | 835 | 110 |