Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nle1
Entrez Gene ID
217011
Gene Name
notchless homolog 1 (Drosophila)
Gene Aliases
AL022765, BC018399, Nle, l11Jus1, l11Jus4
Chromosome Location
Chr.11: 82900768 - 82908445 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145431.2 NP_663406.2 7 - 8 865 69
GenBank mRNA AK141230.1 - 7 - 8 840 69
AK170853.1 - 7 - 8 838 69
BC018399.1 - 7 - 8 856 69
BC035421.1 - 7 - 8 856 69

View Full Product Details