The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145482.3 | NP_663457.2 | 7 - 8 | 1143 | 73 |
GenBank mRNA | AK184278.1 | - | 1 - 2 | 57 | 73 |
AY037804.1 | - | 7 - 8 | 1143 | 73 |