Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc13
Entrez Gene ID
234875
Gene Name
tetratricopeptide repeat domain 13
Gene Aliases
BC017545
Chromosome Location
Chr.8: 124671327 - 124722013 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145607.3 NP_663582.2 14 - 15 1674 78
GenBank mRNA AK144842.1 - 14 - 15 1675 78
AK160894.1 - 16 - 17 1911 78
BC017545.1 - 8 - 9 885 78
BC072577.1 - 14 - 15 1664 78
BC079851.1 - 15 - 16 1729 78

View Full Product Details