The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177706.1 | NP_001171177.1 | 8 - 9 | 1199 | 68 |
NM_001177707.1 | NP_001171178.1 | 9 - 10 | 1070 | 68 | |
NM_001177708.1 | NP_001171179.1 | 8 - 9 | 789 | 68 | |
NM_001177709.1 | NP_001171180.1 | 8 - 9 | 789 | 68 | |
NM_001177710.1 | NP_001171181.1 | 7 - 8 | 679 | 68 | |
NM_145760.3 | NP_665703.2 | 9 - 10 | 1002 | 68 | |
GenBank mRNA | AJ401461.1 | - | 9 - 10 | 960 | 68 |
AK030295.1 | - | 9 - 10 | 958 | 68 | |
AK030520.1 | - | 8 - 9 | 1201 | 68 | |
AK134402.1 | - | 9 - 10 | 1012 | 68 | |
AK145879.1 | - | 9 - 10 | 986 | 68 | |
AK168007.1 | - | 9 - 10 | 983 | 68 | |
AK168602.1 | - | 8 - 9 | 770 | 68 | |
AK216248.1 | - | 3 - 4 | 210 | 68 | |
BC016228.1 | - | 7 - 8 | 637 | 68 | |
BC027794.1 | - | 7 - 8 | 2186 | 68 | |
BC052922.1 | - | 8 - 9 | 737 | 68 | |
BC059817.1 | - | 8 - 9 | 728 | 68 |