The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177706.1 | NP_001171177.1 | 9 - 10 | 1239 | 90 |
NM_001177707.1 | NP_001171178.1 | 10 - 11 | 1110 | 90 | |
NM_001177708.1 | NP_001171179.1 | 9 - 10 | 829 | 90 | |
NM_001177709.1 | NP_001171180.1 | 9 - 10 | 829 | 90 | |
NM_001177710.1 | NP_001171181.1 | 8 - 9 | 719 | 90 | |
NM_145760.3 | NP_665703.2 | 10 - 11 | 1042 | 90 | |
GenBank mRNA | AJ401461.1 | - | 10 - 11 | 1000 | 90 |
AK030295.1 | - | 10 - 11 | 998 | 90 | |
AK030520.1 | - | 9 - 10 | 1241 | 90 | |
AK134402.1 | - | 10 - 11 | 1052 | 90 | |
AK145879.1 | - | 10 - 11 | 1026 | 90 | |
AK168007.1 | - | 10 - 11 | 1023 | 90 | |
AK168602.1 | - | 9 - 10 | 810 | 90 | |
AK216248.1 | - | 4 - 5 | 250 | 90 | |
BC016228.1 | - | 8 - 9 | 677 | 90 | |
BC027794.1 | - | 8 - 9 | 2226 | 90 | |
BC052922.1 | - | 9 - 10 | 777 | 91 | |
BC059817.1 | - | 9 - 10 | 768 | 90 |