The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146087.2 | NP_666199.1 | 2 - 3 | 390 | 122 |
GenBank mRNA | AK076070.1 | - | 2 - 3 | 657 | 122 |
AK076264.1 | - | 2 - 3 | 651 | 122 | |
AK078398.1 | - | 2 - 3 | 639 | 122 | |
AK141421.1 | - | 2 - 3 | 644 | 122 | |
AK146873.1 | - | 2 - 3 | 660 | 122 | |
AK147165.1 | - | 2 - 3 | 635 | 122 | |
AK152387.1 | - | 2 - 3 | 635 | 122 | |
AK152793.1 | - | 2 - 3 | 655 | 122 | |
AK166318.1 | - | 2 - 3 | 650 | 122 | |
AK210280.1 | - | 2 - 3 | 161 | 122 | |
BC019740.1 | - | 2 - 3 | 390 | 122 | |
BC025439.1 | - | 2 - 3 | 337 | 122 | |
BC048081.1 | - | 2 - 3 | 423 | 122 |