Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdc5l
Entrez Gene ID
71702
Gene Name
cell division cycle 5-like (S. pombe)
Gene Aliases
1200002I02Rik, AA408004, PCDC5RP
Chromosome Location
Chr.17: 45391887 - 45433707 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152810.2 NP_690023.1 3 - 4 583 106
GenBank mRNA AK004547.2 - 3 - 4 613 106
AK146892.1 - 3 - 4 403 106
AK150562.1 - 3 - 4 400 106
AK150861.1 - 3 - 4 400 106
AK172950.1 - 3 - 4 403 106
BC003893.1 - 3 - 4 408 106
BC031480.1 - 3 - 4 583 106

View Full Product Details