Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ldb3
Entrez Gene ID
24131
Gene Name
LIM domain binding 3
Gene Aliases
AW742271, PDLIM6, ZASP
Chromosome Location
Chr.14: 34526699 - 34588698 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039071.2 NP_001034160.1 1 - 2 81 62
NM_001039072.2 NP_001034161.1 1 - 2 81 62
NM_001039073.2 NP_001034162.1 1 - 2 81 62
NM_001039075.2 NP_001034164.1 1 - 2 81 62
NM_001039076.2 NP_001034165.1 1 - 2 81 62
NM_011918.4 NP_036048.3 1 - 2 81 62
GenBank mRNA AF114378.2 - 1 - 2 47 62
AF114379.2 - 1 - 2 45 62
AJ005621.1 - 1 - 2 74 62
AK004020.1 - 1 - 2 89 62
AK084461.1 - 1 - 2 61 62
AK137181.1 - 1 - 2 64 62
AK142292.1 - 1 - 2 33 62
AY206012.1 - 1 - 2 47 62
AY206013.1 - 1 - 2 45 62
BC099596.1 - 1 - 2 48 62
BY704269.1 - 1 - 2 88 62

View Full Product Details