Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lman1
Entrez Gene ID
70361
Gene Name
lectin, mannose-binding, 1
Gene Aliases
2610020P13Rik, AI326273, AU043785, C730041J05, ERGIC53, F5F8D, MCFD1, MR60, P58, gp58
Chromosome Location
Chr.18: 65980739 - 66002635 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172062.1 NP_001165533.1 3 - 4 518 134
NM_027400.3 NP_081676.1 3 - 4 518 134
GenBank mRNA AK011495.1 - 3 - 4 504 134
AK151599.1 - 3 - 4 505 134
AK151949.1 - 3 - 4 505 134
AK153554.1 - 3 - 4 505 134
BC057165.1 - 3 - 4 527 134

View Full Product Details