Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcoln1
Entrez Gene ID
94178
Gene Name
mucolipin 1
Gene Aliases
2210015I05Rik, TRPML1, mucolipidin
Chromosome Location
Chr.8: 3500442 - 3515232 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053177.1 NP_444407.1 2 - 3 313 64
GenBank mRNA AF302009.1 - 2 - 3 310 64
AK028385.1 - 2 - 3 290 64
AK034471.1 - 2 - 3 324 64
AK038594.1 - 2 - 3 326 64
AK049606.1 - 2 - 3 314 64
BC005651.1 - 2 - 3 313 64
BC020377.1 - 1 - 2 163 64

View Full Product Details