Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mcoln1
Entrez Gene ID
94178
Gene Name
mucolipin 1
Gene Aliases
2210015I05Rik, TRPML1, mucolipidin
Chromosome Location
Chr.8: 3500442 - 3515232 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053177.1 NP_444407.1 9 - 10 1206 125
GenBank mRNA AF302009.1 - 9 - 10 1203 125
AK028385.1 - 9 - 10 1183 125
AK034471.1 - 9 - 10 1217 125
AK038594.1 - 9 - 10 1219 125
AK049606.1 - 9 - 10 1207 125
BC005651.1 - 9 - 10 1206 125

View Full Product Details