Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mavs
Entrez Gene ID
228607
Gene Name
mitochondrial antiviral signaling protein
Gene Aliases
D430028G21Rik, IPS-1, Visa, cardif
Chromosome Location
Chr.2: 131234063 - 131248025 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206382.1 NP_001193311.1 4 - 5 506 83
NM_001206383.1 NP_001193312.1 2 - 3 240 83
NM_001206385.1 NP_001193314.1 3 - 4 415 83
NM_144888.2 NP_659137.1 3 - 4 643 83
GenBank mRNA AK028421.1 - 3 - 4 414 83
AK085038.1 - 3 - 4 602 83
AK153748.1 - 4 - 5 508 83
BC020006.1 - 3 - 4 375 83
BC025825.1 - 3 - 4 568 83
BC031352.1 - 3 - 4 552 83
BC037391.1 - 2 - 3 354 83
DQ167127.1 - 2 - 3 292 83
DQ174271.1 - 2 - 3 292 83

View Full Product Details