Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp1a3
Entrez Gene ID
232975
Gene Name
ATPase, Na+/K+ transporting, alpha 3 polypeptide
Gene Aliases
Atpa-2
Chromosome Location
Chr.7: 24978167 - 25005937 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290469.1 NP_001277398.1 11 - 12 1612 67
GenBank mRNA BC020177.1 - 11 - 12 1570 67
BC027000.1 - 4 - 5 632 67
BC034645.1 - 11 - 12 1568 67
BC037206.1 - 11 - 12 1567 67
BC042894.1 - 11 - 12 1612 67

View Full Product Details