Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Blvrb
Entrez Gene ID
233016
Gene Name
biliverdin reductase B (flavin reductase (NADPH))
Gene Aliases
-
Chromosome Location
Chr.7: 27447978 - 27465981 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290525.1 NP_001277454.1 2 - 3 297 69
NM_144923.3 NP_659172.1 2 - 3 383 69
GenBank mRNA AK153153.1 - 2 - 3 304 69
BC006617.1 - 2 - 3 323 69
BC027279.1 - 2 - 3 383 69

View Full Product Details