Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lgi2
Entrez Gene ID
246316
Gene Name
leucine-rich repeat LGI family, member 2
Gene Aliases
mKIAA1916
Chromosome Location
Chr.5: 52533517 - 52566462 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144945.2 NP_659194.1 3 - 4 506 89
GenBank mRNA AJ487515.1 - 3 - 4 529 89
AK122570.1 - 3 - 4 471 89
AY841361.1 - 3 - 4 331 89

View Full Product Details