The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285997.1 | NP_001272926.1 | 10 - 11 | 1478 | 74 |
NM_001285998.1 | NP_001272927.1 | 10 - 11 | 1478 | 74 | |
NM_145150.3 | NP_660132.2 | 10 - 11 | 1478 | 74 | |
GenBank mRNA | AF408435.1 | - | 2 - 3 | 237 | 74 |
AK029140.1 | - | 10 - 11 | 1465 | 74 | |
AK088576.1 | - | 10 - 11 | 1450 | 74 | |
AK135778.1 | - | 8 - 9 | 1104 | 74 | |
BC005475.1 | - | 10 - 11 | 1412 | 74 | |
BC059808.1 | - | 10 - 11 | 1423 | 74 |