The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177770.1 | NP_001171241.1 | 10 - 11 | 1741 | 71 |
NM_001177771.1 | NP_001171242.1 | 11 - 12 | 1656 | 71 | |
NM_145543.2 | NP_663518.1 | 10 - 11 | 1453 | 71 | |
GenBank mRNA | AK143631.1 | - | 11 - 12 | 1640 | 71 |
AK169803.1 | - | 10 - 11 | 1451 | 71 | |
AK171296.1 | - | 10 - 11 | 1480 | 71 | |
BC003247.1 | - | 10 - 11 | 1484 | 71 |