The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145584.2 | NP_663559.1 | 2 - 3 | 678 | 91 |
GenBank mRNA | AK134774.1 | - | 2 - 3 | 503 | 91 |
AK146506.1 | - | 2 - 3 | 503 | 91 | |
AK173021.1 | - | 2 - 3 | 345 | 91 | |
BC020531.1 | - | 2 - 3 | 548 | 91 |