Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serinc4
Entrez Gene ID
574418
Gene Name
serine incorporator 4
Gene Aliases
-
Chromosome Location
Chr.2: 121451177 - 121456764 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025371.2 NP_001020542.1 5 - 6 829 81
GenBank mRNA AK044479.1 - 5 - 6 857 81
BC026459.1 - 5 - 6 758 81
BC083132.1 - 5 - 6 829 81

View Full Product Details