The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145974.3 | NP_666086.2 | 2 - 3 | 745 | 88 |
GenBank mRNA | AK049249.1 | - | 2 - 3 | 515 | 88 |
AK077848.1 | - | 2 - 3 | 508 | 88 | |
AK141228.1 | - | 2 - 3 | 539 | 88 | |
AK142500.1 | - | 2 - 3 | 508 | 88 | |
AK170270.1 | - | 2 - 3 | 543 | 88 | |
AK170521.1 | - | 2 - 3 | 540 | 88 | |
BC006740.1 | - | 2 - 3 | 458 | 88 | |
BC055831.1 | - | 2 - 3 | 745 | 88 |