The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253736.1 | NP_001240665.1 | 8 - 9 | 893 | 65 |
NM_145978.2 | NP_666090.1 | 8 - 9 | 820 | 65 | |
GenBank mRNA | BC024556.1 | - | 8 - 9 | 820 | 65 |
BE634101.1 | - | 4 - 5 | 315 | 65 |