The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289711.1 | NP_001276640.1 | 12 - 13 | 1615 | 70 |
NM_001289712.1 | NP_001276641.1 | 12 - 13 | 1615 | 70 | |
NM_145993.5 | NP_666105.2 | 12 - 13 | 1615 | 70 | |
NR_110362.1 | 12 - 13 | 1938 | 70 | ||
GenBank mRNA | AK029115.1 | - | 12 - 13 | 1560 | 70 |
AK036510.1 | - | 12 - 13 | 1568 | 70 | |
AK132012.1 | - | 12 - 13 | 1567 | 70 | |
AK141810.1 | - | 11 - 12 | 2748 | 70 | |
AK145560.1 | - | 12 - 13 | 1557 | 70 | |
AK146632.1 | - | 12 - 13 | 1571 | 70 | |
AK156236.1 | - | 12 - 13 | 1570 | 70 | |
AK211064.1 | - | 2 - 3 | 253 | 70 | |
AY237000.1 | - | 12 - 13 | 1548 | 70 | |
AY237001.1 | - | 12 - 13 | 1548 | 70 | |
AY237002.1 | - | 12 - 13 | 1871 | 70 | |
BC023676.1 | - | 1 - 2 | 150 | 70 | |
BC023933.1 | - | 12 - 13 | 1556 | 70 | |
BC030864.1 | - | 12 - 13 | 1518 | 70 |