The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146040.1 | NP_666152.1 | 9 - 10 | 1378 | 85 |
GenBank mRNA | AK149673.1 | - | 9 - 10 | 1356 | 85 |
BC006933.1 | - | 9 - 10 | 1378 | 85 | |
BC071198.1 | - | 9 - 10 | 1346 | 85 |