Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubxn1
Entrez Gene ID
225896
Gene Name
UBX domain protein 1
Gene Aliases
2B28, 4930455J02Rik, D19Ertd721e, T25529
Chromosome Location
Chr.19: 8871510 - 8875664 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146093.1 NP_666205.1 4 - 5 277 70
GenBank mRNA AK015468.1 - 3 - 4 329 70
AK150442.1 - 3 - 4 280 70
AK159708.1 - 3 - 4 253 70
BC006701.1 - 4 - 5 277 70

View Full Product Details