Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gimap7
Entrez Gene ID
231932
Gene Name
GTPase, IMAP family member 7
Gene Aliases
IAN7, Ian3
Chromosome Location
Chr.6: 48718621 - 48724636 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146167.3 NP_666279.1 1 - 2 81 63
GenBank mRNA AA197670.1 - 1 - 2 59 63
AK040029.1 - 1 - 2 40 63
BC026200.1 - 1 - 2 43 63
BE448872.1 - 1 - 2 81 63

View Full Product Details