Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fyco1
Entrez Gene ID
17281
Gene Name
FYVE and coiled-coil domain containing 1
Gene Aliases
2810409M01Rik, Mem2, RUFY3, ZFYVE7
Chromosome Location
Chr.9: 123789500 - 123851899 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110253.2 NP_001103723.2 9 - 10 3235 78
NM_148925.3 NP_683727.3 10 - 11 3482 78
GenBank mRNA AJ428065.1 - 10 - 11 3387 78
AK044587.1 - 2 - 3 544 78
CB244718.1 - 1 - 2 68 78

View Full Product Details