Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ap1b1
Entrez Gene ID
11764
Gene Name
adaptor protein complex AP-1, beta 1 subunit
Gene Aliases
Adtb1, b2b1660Clo
Chromosome Location
Chr.11: 4947521 - 5042794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243043.1 NP_001229972.1 19 - 20 2989 78
NM_001243044.1 NP_001229973.1 18 - 19 2771 78
NM_007454.3 NP_031480.2 18 - 19 2771 78
GenBank mRNA AA003413.1 - 4 - 5 407 78
AK034140.1 - 19 - 20 2992 78
AK155892.1 - 18 - 19 2691 78
AK159273.1 - 18 - 19 2662 78
AK160042.1 - 18 - 19 2662 78
AK165747.1 - 18 - 19 2690 78
AK170376.1 - 18 - 19 2682 78
BC008513.1 - 18 - 19 2584 78
BC066827.1 - 9 - 10 1315 78
Y07919.1 - 18 - 19 2626 78

View Full Product Details