Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aplp1
Entrez Gene ID
11803
Gene Name
amyloid beta (A4) precursor-like protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 30434980 - 30445582 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007467.3 NP_031493.2 7 - 8 1109 63
GenBank mRNA AK154992.1 - 7 - 8 1063 63
AK160871.1 - 7 - 8 1061 63
AK170818.1 - 7 - 8 1042 63
BC021877.1 - 7 - 8 1088 63
BC106158.1 - 6 - 7 1197 63
L04538.1 - 7 - 8 1079 63

View Full Product Details