The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164627.1 | NP_001158099.1 | 8 - 9 | 712 | 102 |
NM_001164628.1 | NP_001158100.1 | 7 - 8 | 704 | 102 | |
NM_001205334.1 | NP_001192263.1 | 8 - 9 | 732 | 102 | |
NM_028455.4 | NP_082731.2 | 7 - 8 | 724 | 102 | |
GenBank mRNA | AK014171.1 | - | 7 - 8 | 675 | 102 |
AK086816.1 | - | 7 - 8 | 678 | 102 | |
AK088242.1 | - | 7 - 8 | 680 | 102 | |
BC005563.1 | - | 6 - 7 | 667 | 102 | |
BC010306.1 | - | 7 - 8 | 694 | 102 |