Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgap12
Entrez Gene ID
75415
Gene Name
Rho GTPase activating protein 12
Gene Aliases
2810011M08Rik
Chromosome Location
Chr.18: 6024448 - 6136098 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039692.1 NP_001034781.1 5 - 6 1492 100
NM_029277.2 NP_083553.2 5 - 6 1492 100
GenBank mRNA AK012717.2 - 5 - 6 1334 100
AK031648.1 - 5 - 6 1336 100
AK077063.1 - 5 - 6 1493 100
BC037608.1 - 5 - 6 1307 100
BC141184.1 - 5 - 6 1301 100
BC145506.1 - 5 - 6 1300 100

View Full Product Details