Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crispld2
Entrez Gene ID
78892
Gene Name
cysteine-rich secretory protein LCCL domain containing 2
Gene Aliases
1810049K24Rik, Lcrisp2, Lgl1, coffeecrisp
Chromosome Location
Chr.8: 119992443 - 120052793 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030209.3 NP_084485.1 11 - 12 1363 91
GenBank mRNA AK019034.1 - 11 - 12 1359 91
AK033858.1 - 11 - 12 1327 91
AK132171.1 - 11 - 12 1306 91
AK149909.1 - 11 - 12 1320 91
AK149925.1 - 11 - 12 1320 91
AK150180.1 - 11 - 12 1320 91
AK154308.1 - 11 - 12 1363 91
BC090642.1 - 11 - 12 1357 91

View Full Product Details