Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Entpd6
- Entrez Gene ID
-
12497
- Gene Name
- ectonucleoside triphosphate diphosphohydrolase 6
- Gene Aliases
-
2700026H11Rik, Cd39l2, NTPDase-6
- Chromosome Location
-
Chr.2: 150749038 - 150771675 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC038126.1 |
- |
7 - 8 |
889 |
138 |
View Full Product Details