Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Noxa1
Entrez Gene ID
241275
Gene Name
NADPH oxidase activator 1
Gene Aliases
NY-CO-31, SDCCAG31
Chromosome Location
Chr.2: 25085667 - 25095205 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163626.1 NP_001157098.1 1 - 2 462 78
NM_172204.4 NP_757341.3 1 - 2 462 78
GenBank mRNA AF539798.1 - 1 - 2 181 78
AK087263.1 - 1 - 2 388 78
AY255770.1 - 1 - 2 181 78
BC047532.1 - 1 - 2 453 78

View Full Product Details