The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009703.2 | NP_033833.1 | 10 - 11 | 1210 | 85 |
GenBank mRNA | AK085823.1 | - | 10 - 11 | 1176 | 85 |
BC004757.1 | - | 10 - 11 | 1166 | 85 | |
D00024.1 | - | 5 - 6 | 572 | 85 |