Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dctn2
Entrez Gene ID
69654
Gene Name
dynactin 2
Gene Aliases
2310042E05Rik, C130077D06Rik, C87049, DCTN-50, GMP23-48K, RBP50, p50
Chromosome Location
Chr.10: 127266262 - 127281959 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190453.1 NP_001177382.1 12 - 13 1172 57
NM_001190454.1 NP_001177383.1 11 - 12 1181 57
NM_027151.2 NP_081427.1 11 - 12 1166 57
GenBank mRNA AK009749.1 - 6 - 7 418 57
AK144464.1 - 11 - 12 1042 57
AK144505.1 - 11 - 12 1043 57
AK164021.1 - 11 - 12 1043 57
BC004613.1 - 11 - 12 1029 57

View Full Product Details