The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027592.3 | NP_081868.1 | 2 - 3 | 198 | 150 |
GenBank mRNA | BC019453.1 | - | 2 - 3 | 149 | 150 |