Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Txndc11
Entrez Gene ID
106200
Gene Name
thioredoxin domain containing 11
Gene Aliases
2810408E11Rik, AI427833, EFP1, Txdc11
Chromosome Location
Chr.16: 11074911 - 11134681 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029582.2 NP_083858.1 3 - 4 587 123
GenBank mRNA AK013044.1 - 3 - 4 635 123
AK029166.1 - 3 - 4 706 123
AK139620.1 - 3 - 4 669 123
AK155844.1 - 3 - 4 641 123
AK156154.1 - 3 - 4 628 123
AK156590.1 - 3 - 4 667 123
AK157288.1 - 3 - 4 561 123
AK157948.1 - 3 - 4 667 123
AK159484.1 - 3 - 4 644 123
AK170352.1 - 3 - 4 628 123
BC020363.1 - 3 - 4 676 123
BC029643.1 - 3 - 4 587 123

View Full Product Details