Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Txndc11
Entrez Gene ID
106200
Gene Name
thioredoxin domain containing 11
Gene Aliases
2810408E11Rik, AI427833, EFP1, Txdc11
Chromosome Location
Chr.16: 11074911 - 11134681 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029582.2 NP_083858.1 4 - 5 707 81
GenBank mRNA AK013044.1 - 4 - 5 755 81
AK139620.1 - 4 - 5 789 81
AK155844.1 - 4 - 5 761 81
AK156154.1 - 4 - 5 748 81
AK157288.1 - 4 - 5 681 81
AK157948.1 - 4 - 5 787 81
AK159484.1 - 4 - 5 764 81
AK170352.1 - 4 - 5 748 81
BC020363.1 - 4 - 5 796 81
BC029643.1 - 4 - 5 707 81

View Full Product Details