Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Upp2
Entrez Gene ID
76654
Gene Name
uridine phosphorylase 2
Gene Aliases
1700124F02Rik, AI266885, UDRPASE2, UP2, UPASE2
Chromosome Location
Chr.2: 58567225 - 58793042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289659.1 NP_001276588.1 7 - 8 1048 74
NM_029692.3 NP_083968.1 6 - 7 994 74
GenBank mRNA AK007264.1 - 5 - 6 671 74
AK149444.1 - 6 - 7 992 74
AY152393.1 - 6 - 7 993 74
BC027189.1 - 7 - 8 1290 74

View Full Product Details