Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fuk
Entrez Gene ID
234730
Gene Name
fucokinase
Gene Aliases
1110046B12Rik, AI303278
Chromosome Location
Chr.8: 110882456 - 110902488 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172283.3 NP_758487.2 14 - 15 1496 113
NR_052009.1 14 - 15 1594 113
GenBank mRNA AJ297482.4 - 14 - 15 1467 113
AJ534942.1 - 14 - 15 1467 113
AK049153.1 - 3 - 4 325 113
AK161763.1 - 14 - 15 1598 113
AK164340.1 - 14 - 15 1563 113
AY223865.1 - 14 - 15 1467 113
BC037698.1 - 13 - 14 1445 113

View Full Product Details