Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc36a4
Entrez Gene ID
234967
Gene Name
solute carrier family 36 (proton/amino acid symporter), member 4
Gene Aliases
6330573I15Rik, PAT4
Chromosome Location
Chr.9: 15709769 - 15742388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172289.4 NP_758493.2 9 - 10 1045 117
GenBank mRNA AF453746.1 - 10 - 11 1044 117
AK032074.1 - 9 - 10 1046 117
AK052277.1 - 9 - 10 1046 117
AK150167.1 - 9 - 10 1077 117
BC115964.1 - 9 - 10 1046 117

View Full Product Details