The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083331.2 | NP_001076800.1 | 2 - 3 | 491 | 94 |
NM_001276406.1 | NP_001263335.1 | 2 - 3 | 491 | 94 | |
NM_172394.3 | NP_765982.2 | 2 - 3 | 491 | 94 | |
NR_075087.1 | 2 - 3 | 1053 | 94 | ||
GenBank mRNA | AJ532593.1 | - | 2 - 3 | 485 | 94 |
AK002643.1 | - | 2 - 3 | 477 | 94 | |
AK028563.1 | - | 2 - 3 | 481 | 94 | |
AK039007.1 | - | 2 - 3 | 481 | 94 | |
AK050893.1 | - | 2 - 3 | 480 | 94 | |
AK085776.1 | - | 2 - 3 | 492 | 94 | |
AK139738.1 | - | 2 - 3 | 961 | 94 | |
AK151739.1 | - | 2 - 3 | 481 | 94 | |
AK160619.1 | - | 2 - 3 | 492 | 94 | |
AK166214.1 | - | 2 - 3 | 481 | 94 | |
BC032929.1 | - | 2 - 3 | 472 | 94 |