The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161767.1 | NP_001155239.1 | 11 - 12 | 2148 | 67 |
NM_001161768.1 | NP_001155240.1 | 11 - 12 | 2145 | 67 | |
NM_172451.3 | NP_766039.2 | 11 - 12 | 2144 | 67 | |
GenBank mRNA | AJ133523.1 | - | 9 - 10 | 1760 | 67 |
AK028506.1 | - | 11 - 12 | 2144 | 67 | |
AK049222.1 | - | 11 - 12 | 2147 | 67 | |
AK155008.1 | - | 11 - 12 | 2151 | 67 | |
AK159721.1 | - | 11 - 12 | 2126 | 67 | |
BC119324.1 | - | 9 - 10 | 1797 | 67 | |
BC119326.1 | - | 9 - 10 | 1797 | 67 |