The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172461.3 | NP_766049.2 | 12 - 13 | 1521 | 115 |
GenBank mRNA | AK030042.1 | - | 12 - 13 | 1480 | 115 |
AK030186.1 | - | 11 - 12 | 1397 | 115 | |
BC119588.1 | - | 11 - 12 | 1355 | 115 | |
BC119589.1 | - | 11 - 12 | 1355 | 115 |