The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159527.1 | NP_001152999.1 | 26 - 27 | 3522 | 76 |
NM_172470.3 | NP_766058.3 | 27 - 28 | 3555 | 76 | |
GenBank mRNA | AK029896.1 | - | 26 - 27 | 3477 | 76 |
AK083971.1 | - | 26 - 27 | 3467 | 76 | |
BC056925.1 | - | 27 - 28 | 3664 | 76 |