Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Itih5
Entrez Gene ID
209378
Gene Name
inter-alpha (globulin) inhibitor H5
Gene Aliases
4631408O11Rik, 5430408M01Rik, E130106B02
Chromosome Location
Chr.2: 10153536 - 10256529 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172471.2 NP_766059.1 13 - 14 2655 70
GenBank mRNA AK087428.1 - 13 - 14 2583 70
AK134707.1 - 13 - 14 2583 70
AK140756.1 - 13 - 14 2656 70
BC043314.1 - 13 - 14 2598 70
BC062196.1 - 13 - 14 2639 70

View Full Product Details