Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hace1
Entrez Gene ID
209462
Gene Name
HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1
Gene Aliases
1700042J16Rik, A730034A22Rik, BC025474
Chromosome Location
Chr.10: 45577571 - 45712345 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172473.3 NP_766061.2 22 - 23 2757 109
GenBank mRNA AK029162.1 - 12 - 13 1577 109
AK042879.1 - 22 - 23 2698 109
AK156958.1 - 22 - 23 2699 109
BC025227.1 - 6 - 7 682 109
BC120695.1 - 22 - 23 2654 109
BC120697.1 - 22 - 23 2654 109

View Full Product Details