The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172476.4 | NP_766064.2 | 7 - 8 | 1018 | 79 |
GenBank mRNA | AK082562.1 | - | 7 - 8 | 1047 | 79 |
AK083189.1 | - | 7 - 8 | 1021 | 79 | |
AK132820.1 | - | 7 - 8 | 1047 | 79 | |
AY236499.1 | - | 7 - 8 | 1079 | 79 | |
AY263161.1 | - | 7 - 8 | 1047 | 79 | |
BC105966.1 | - | 1 - 2 | 49 | 79 |